Assay Details
| Gene Symbol: Adam12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007400.2 | NP_031426.2 | 21 - 22 | 2743 | 55 | |
| GenBank mRNA | AK051780.1 | - | 11 - 12 | 1557 | 55 | |
| AK220274.1 | - | 8 - 9 | 953 | 55 | ||
| D50411.1 | - | 21 - 22 | 2743 | 55 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Adam12
Genomic Map