Assay Details
| Gene Symbol: Tbc1d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285991.1 | NP_001272920.1 | 21 - 22 | 2425 | 133 | |
| NM_001285993.1 | NP_001272922.1 | 20 - 21 | 2326 | 133 | ||
| NM_028162.4 | NP_082438.3 | 20 - 21 | 2359 | 133 | ||
| GenBank mRNA | AK154975.1 | - | 21 - 22 | 2385 | 133 | |
| AK163366.1 | - | 20 - 21 | 2389 | 133 | ||
| AK220251.1 | - | 20 - 21 | 2326 | 133 | ||
| BC043113.1 | - | 20 - 21 | 2175 | 6407234 | 133 | |
| BC098328.1 | - | 15 - 16 | 1705 | 30357143 | 133 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d5
Genomic Map