Assay Details
| Gene Symbol: Tbc1d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285991.1 | NP_001272920.1 | 8 - 9 | 858 | 118 | |
| NM_001285993.1 | NP_001272922.1 | 8 - 9 | 825 | 118 | ||
| NM_028162.4 | NP_082438.3 | 8 - 9 | 858 | 118 | ||
| GenBank mRNA | AK005461.1 | - | 8 - 9 | 842 | 118 | |
| AK033710.1 | - | 8 - 9 | 750 | 118 | ||
| AK144467.1 | - | 8 - 9 | 859 | 118 | ||
| AK154975.1 | - | 8 - 9 | 818 | 118 | ||
| AK163366.1 | - | 8 - 9 | 833 | 119 | ||
| AK220251.1 | - | 8 - 9 | 825 | 118 | ||
| BC043113.1 | - | 8 - 9 | 674 | 6407234 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d5
Genomic Map