Assay Details
| Gene Symbol: Ascl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008554.3 | NP_032580.2 | 1 - 2 | 294 | 67 | |
| GenBank mRNA | AK010738.1 | - | 1 - 2 | 259 | 67 | |
| BC019520.1 | - | 1 - 2 | 269 | 4163833 | 67 | |
| CJ074389.1 | - | 1 - 2 | 295 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC019520.1, NM_008554.3 transcripts.
Set Membership: |
|

Ascl2
Genomic Map