Assay Details
| Gene Symbol: Sec23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252543.1 | NP_001239472.1 | 3 - 4 | 398 | 69 | |
| NM_001252544.1 | NP_001239473.1 | 3 - 4 | 391 | 69 | ||
| NM_001252545.1 | NP_001239474.1 | 3 - 4 | 380 | 69 | ||
| NM_019787.4 | NP_062761.2 | 3 - 4 | 409 | 69 | ||
| GenBank mRNA | AF200326.1 | - | 2 - 3 | 281 | 69 | |
| AK014597.1 | - | 3 - 4 | 359 | 69 | ||
| AK139729.1 | - | 3 - 4 | 323 | 69 | ||
| AK146083.1 | - | 3 - 4 | 358 | 69 | ||
| AK149693.1 | - | 3 - 4 | 338 | 69 | ||
| AK153214.1 | - | 3 - 4 | 365 | 69 | ||
| AK155168.1 | - | 3 - 4 | 339 | 69 | ||
| AK158398.1 | - | 3 - 4 | 376 | 69 | ||
| AK166330.1 | - | 3 - 4 | 341 | 69 | ||
| AK167382.1 | - | 3 - 4 | 347 | 69 | ||
| AK167732.1 | - | 3 - 4 | 327 | 69 | ||
| AK171597.1 | - | 3 - 4 | 339 | 69 | ||
| AK179166.1 | - | 2 - 3 | 201 | 69 | ||
| BC005464.1 | - | 3 - 4 | 334 | 3591420 | 69 | |
| BC011160.1 | - | 3 - 4 | 361 | 4024353 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sec23b
Genomic Map