Assay Details
| Gene Symbol: Sec23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252543.1 | NP_001239472.1 | 5 - 6 | 722 | 67 | |
| NM_001252544.1 | NP_001239473.1 | 5 - 6 | 715 | 67 | ||
| NM_001252545.1 | NP_001239474.1 | 5 - 6 | 704 | 67 | ||
| NM_001281816.1 | NP_001268745.1 | 2 - 3 | 344 | 67 | ||
| NM_019787.4 | NP_062761.2 | 5 - 6 | 733 | 67 | ||
| GenBank mRNA | AF200326.1 | - | 4 - 5 | 605 | 67 | |
| AK014597.1 | - | 5 - 6 | 683 | 67 | ||
| AK029661.1 | - | 2 - 3 | 345 | 67 | ||
| AK139729.1 | - | 5 - 6 | 647 | 67 | ||
| AK146083.1 | - | 5 - 6 | 682 | 67 | ||
| AK149693.1 | - | 5 - 6 | 662 | 67 | ||
| AK153214.1 | - | 5 - 6 | 689 | 67 | ||
| AK155168.1 | - | 5 - 6 | 663 | 67 | ||
| AK158398.1 | - | 5 - 6 | 700 | 67 | ||
| AK166330.1 | - | 5 - 6 | 665 | 67 | ||
| AK167382.1 | - | 5 - 6 | 671 | 67 | ||
| AK167732.1 | - | 5 - 6 | 651 | 67 | ||
| AK171597.1 | - | 5 - 6 | 663 | 67 | ||
| BC005464.1 | - | 5 - 6 | 658 | 3591420 | 67 | |
| BC011160.1 | - | 5 - 6 | 685 | 4024353 | 67 | |
| BU614902.1 | - | 1 - 2 | 62 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec23b
Genomic Map