Assay Details
| Gene Symbol: Sec23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252543.1 | NP_001239472.1 | 6 - 7 | 803 | 74 | |
| NM_001252544.1 | NP_001239473.1 | 6 - 7 | 796 | 74 | ||
| NM_001252545.1 | NP_001239474.1 | 6 - 7 | 785 | 74 | ||
| NM_001281816.1 | NP_001268745.1 | 3 - 4 | 425 | 74 | ||
| NM_019787.4 | NP_062761.2 | 6 - 7 | 814 | 74 | ||
| GenBank mRNA | AF200326.1 | - | 5 - 6 | 686 | 74 | |
| AK014597.1 | - | 6 - 7 | 764 | 74 | ||
| AK029661.1 | - | 3 - 4 | 426 | 74 | ||
| AK139729.1 | - | 6 - 7 | 729 | 74 | ||
| AK149693.1 | - | 6 - 7 | 743 | 74 | ||
| AK153214.1 | - | 6 - 7 | 770 | 74 | ||
| AK155168.1 | - | 6 - 7 | 744 | 74 | ||
| AK158398.1 | - | 6 - 7 | 781 | 74 | ||
| AK166330.1 | - | 6 - 7 | 746 | 74 | ||
| AK167382.1 | - | 6 - 7 | 752 | 74 | ||
| AK167732.1 | - | 6 - 7 | 732 | 74 | ||
| AK171597.1 | - | 6 - 7 | 744 | 74 | ||
| AK207187.1 | - | 2 - 3 | 294 | 74 | ||
| BC005464.1 | - | 6 - 7 | 739 | 3591420 | 74 | |
| BC011160.1 | - | 6 - 7 | 766 | 4024353 | 74 | |
| BU614902.1 | - | 2 - 3 | 143 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec23b
Genomic Map