Assay Details
| Gene Symbol: Cyp39a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285947.1 | NP_001272876.1 | 2 - 3 | 560 | 142 | |
| NM_001285948.1 | NP_001272877.1 | 2 - 3 | 560 | 142 | ||
| NM_018887.4 | NP_061375.1 | 2 - 3 | 560 | 142 | ||
| GenBank mRNA | AF237981.1 | - | 2 - 3 | 409 | 142 | |
| AK031733.1 | - | 2 - 3 | 508 | 142 | ||
More Information
Set Membership: |
|

Cyp39a1
Genomic Map