Assay Details
| Gene Symbol: Cyp39a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285948.1 | NP_001272877.1 | 4 - 5 | 880 | 85 | |
| NM_018887.4 | NP_061375.1 | 4 - 5 | 880 | 85 | ||
| GenBank mRNA | AF237981.1 | - | 4 - 5 | 729 | 85 | |
| AK031733.1 | - | 4 - 5 | 828 | 85 | ||
| BC021418.1 | - | 4 - 5 | 751 | 5025117 | 85 | |
More Information
Set Membership: |
|

Cyp39a1
Genomic Map