Assay Details
| Gene Symbol: Cyp39a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285947.1 | NP_001272876.1 | 8 - 9 | 1255 | 135 | |
| NM_018887.4 | NP_061375.1 | 9 - 10 | 1405 | 135 | ||
| GenBank mRNA | AF237981.1 | - | 9 - 10 | 1254 | 135 | |
| AK031733.1 | - | 9 - 10 | 1353 | 135 | ||
| BC037663.1 | - | 8 - 9 | 1104 | 5352740 | 135 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp39a1
Genomic Map