Assay Details
| Gene Symbol: Adgrf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081178.1 | NP_001074647.1 | 13 - 14 | 2101 | 57 | |
| GenBank mRNA | AK173020.1 | - | 7 - 8 | 1220 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adgrf5
Genomic Map