Assay Details
| Gene Symbol: Adgrf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081178.1 | NP_001074647.1 | 17 - 18 | 4046 | 83 | |
| GenBank mRNA | AK034382.1 | - | 1 - 2 | 82 | 83 | |
| AK173020.1 | - | 11 - 12 | 3165 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adgrf5
Genomic Map