Assay Details
| Gene Symbol: Gcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF276754.1 | - | 2 - 3 | 197 | 69 | |
| AK007911.1 | - | 2 - 3 | 69 | |||
| AK148544.1 | - | 2 - 3 | 191 | 69 | ||
| BC012975.1 | - | 2 - 3 | 192 | 4218718 | 69 | |
| Z46845.1 | - | 1 - 2 | 125 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcg
Genomic Map