Assay Details
| Gene Symbol: Gcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007911.1 | - | 4 - 5 | 484 | 62 | |
| AK148544.1 | - | 4 - 5 | 485 | 62 | ||
| BC012975.1 | - | 4 - 5 | 486 | 4218718 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gcg
Genomic Map