Assay Details
| Gene Symbol: Sphkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019735.1 | - | 4 - 5 | 2047 | 150 | |
| AK122538.1 | - | 4 - 5 | 3876 | 150 | ||
| BC060165.1 | - | 4 - 5 | 2059 | 150 | ||
More Information
Set Membership: |
|

Sphkap
Genomic Map