Assay Details
| Gene Symbol: Sphkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020841.1 | - | 1 - 2 | 122 | 60 | |
| AK034183.1 | - | 1 - 2 | 190 | 60 | ||
| AK038638.1 | - | 1 - 2 | 192 | 60 | ||
More Information
Set Membership: |
|

Sphkap
Genomic Map