Assay Details
| Gene Symbol: Sphkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172430.3 | NP_766018.3 | 5 - 6 | 621 | 71 | |
| GenBank mRNA | AK034183.1 | - | 6 - 7 | 664 | 71 | |
| AK038638.1 | - | 6 - 7 | 666 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sphkap
Genomic Map