Assay Details
| Gene Symbol: Sphkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172430.3 | NP_766018.3 | 6 - 7 | 886 | 117 | |
| GenBank mRNA | AK020841.1 | - | 7 - 8 | 861 | 117 | |
| AK034183.1 | - | 7 - 8 | 929 | 117 | ||
| AK038638.1 | - | 7 - 8 | 931 | 117 | ||
| BC069832.1 | - | 6 - 7 | 852 | 30431760 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sphkap
Genomic Map