Assay Details
| Gene Symbol: Art2a-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF016462.1 | - | 772 | 120 | ||
| BC120753.1 | - | 866 | 40129676 | 120 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Art2a-ps| Gene Symbol: Art2a-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF016462.1 | - | 772 | 120 | ||
| BC120753.1 | - | 866 | 40129676 | 120 | ||
Set Membership: |
|