Assay Details
| Gene Symbol: Mut | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008650.3 | NP_032676.2 | 9 - 10 | 1811 | 107 | |
| GenBank mRNA | AK146309.1 | - | 9 - 10 | 1771 | 107 | |
| BC019175.1 | - | 9 - 10 | 1793 | 5007694 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mut
Genomic Map