Assay Details
| Gene Symbol: Cenpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031863.3 | NP_114069.1 | 5 - 6 | 546 | 127 | |
| GenBank mRNA | AK011998.1 | - | 5 - 6 | 532 | 127 | |
| AK012179.1 | - | 5 - 6 | 547 | 127 | ||
| AK020040.1 | - | 5 - 6 | 548 | 127 | ||
| AK076094.1 | - | 5 - 6 | 524 | 127 | ||
| AK157569.1 | - | 5 - 6 | 571 | 127 | ||
| BC018524.1 | - | 5 - 6 | 479 | 3588374 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenpq
Genomic Map