Assay Details
| Gene Symbol: Cenpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031863.3 | NP_114069.1 | 7 - 8 | 797 | 149 | |
| GenBank mRNA | AK011998.1 | - | 7 - 8 | 783 | 149 | |
| AK012179.1 | - | 7 - 8 | 798 | 149 | ||
| AK076094.1 | - | 7 - 8 | 775 | 149 | ||
| AK157569.1 | - | 7 - 8 | 822 | 149 | ||
| BC018524.1 | - | 7 - 8 | 730 | 3588374 | 149 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenpq
Genomic Map