Assay Details
| Gene Symbol: Ctdspl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290991.1 | NP_001277920.1 | 11 - 12 | 1867 | 158 | |
| NM_001290992.1 | NP_001277921.1 | 11 - 12 | 1866 | 158 | ||
| NM_212450.4 | NP_997615.1 | 11 - 12 | 1870 | 158 | ||
| GenBank mRNA | AK028251.1 | - | 9 - 10 | 930 | 158 | |
| AK030085.1 | - | 11 - 12 | 1386 | 158 | ||
| AK033382.1 | - | 11 - 12 | 1419 | 158 | ||
| AK044805.1 | - | 11 - 12 | 1401 | 158 | ||
| AK049463.1 | - | 11 - 12 | 1386 | 158 | ||
| AK053513.1 | - | 11 - 12 | 1396 | 158 | ||
| AK082646.1 | - | 11 - 12 | 1726 | 158 | ||
| AK163262.1 | - | 11 - 12 | 1871 | 158 | ||
| BC052660.1 | - | 11 - 12 | 1353 | 30049400 | 158 | |
| BC066219.1 | - | 11 - 12 | 1387 | 158 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ctdspl2
Genomic Map