Assay Details
| Gene Symbol: Ctdspl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290991.1 | NP_001277920.1 | 2 - 3 | 826 | 99 | |
| NM_212450.4 | NP_997615.1 | 2 - 3 | 826 | 99 | ||
| GenBank mRNA | AK030085.1 | - | 2 - 3 | 342 | 99 | |
| AK033382.1 | - | 2 - 3 | 375 | 99 | ||
| AK041422.1 | - | 2 - 3 | 342 | 99 | ||
| AK049463.1 | - | 2 - 3 | 342 | 99 | ||
| AK053513.1 | - | 2 - 3 | 355 | 99 | ||
| AK082646.1 | - | 2 - 3 | 682 | 99 | ||
| AK160431.1 | - | 2 - 3 | 341 | 99 | ||
| AK163262.1 | - | 2 - 3 | 827 | 99 | ||
| AK169248.1 | - | 2 - 3 | 368 | 99 | ||
| BC052660.1 | - | 2 - 3 | 309 | 30049400 | 99 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ctdspl2
Genomic Map