Assay Details
| Gene Symbol: Shf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013829.2 | NP_001013851.2 | 4 - 5 | 791 | 60 | |
| GenBank mRNA | BC042839.1 | - | 4 - 5 | 679 | 4488005 | 60 |
| CB056323.1 | - | 3 - 4 | 405 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shf
Genomic Map