Assay Details
| Gene Symbol: Nlrp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002894.2 | NP_001002894.2 | 3 - 4 | 1765 | 89 | |
| GenBank mRNA | AY596199.1 | - | 2 - 3 | 1641 | 89 | |
| AY673647.1 | - | 3 - 4 | 1765 | 89 | ||
| BC141199.1 | - | 3 - 4 | 1749 | 9055811 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nlrp14
Genomic Map