Assay Details
| Gene Symbol: Nlrp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002894.2 | NP_001002894.2 | 5 - 6 | 2101 | 89 | |
| GenBank mRNA | AY596199.1 | - | 4 - 5 | 89 | ||
| AY673647.1 | - | 5 - 6 | 2101 | 89 | ||
| BC141199.1 | - | 5 - 6 | 2085 | 9055811 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nlrp14
Genomic Map