Assay Details
| Gene Symbol: Gltscr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100452.1 | NP_001093922.1 | 10 - 11 | 2863 | 105 | |
| GenBank mRNA | AB093219.1 | - | 7 - 8 | 2016 | 105 | |
| AK029927.1 | - | 11 - 12 | 2508 | 105 | ||
| BC032203.1 | - | 7 - 8 | 1544 | 105 | ||
| BC151032.1 | - | 10 - 11 | 2863 | 9087945 | 105 | |
| BC151054.1 | - | 10 - 11 | 2863 | 9087967 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P290341 | BC058990.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P290341 | BC151032.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P290341 | BC151054.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P290341 | AK029927.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P290341 | NM_001100452.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P290341 | BC058990.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P290341 | BC151032.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P290341 | BC151054.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P290341 | AK029927.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P290341 | NM_001100452.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P122843 | BC058990.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P122843 | AK133487.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P122843 | NM_001100452.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P122843 | AB093219.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P122843 | BC058990.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P122843 | AK133487.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P122843 | NM_001100452.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P122843 | AB093219.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676161 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522026 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410719 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352472 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59208 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868569 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 536282 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 962243 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 683328 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 118856 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 356891 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 245954 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781516 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 948075 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 128328 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 660100 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 175789 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 837061 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 462937 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 678996 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 436588 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578898 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 130704 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 170760 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147755 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 100701 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13286 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101639 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127244 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156219 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143497 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179181 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146610 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109634 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150622 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131334 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166971 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P122843 | BC058990.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P122843 | AK133487.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P122843 | NM_001100452.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P122843 | AB093219.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P122843 | BC058990.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P122843 | AK133487.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P122843 | NM_001100452.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P122843 | AB093219.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P290341 | BC058990.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P290341 | BC151032.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P290341 | BC151054.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P290341 | AK029927.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P290341 | NM_001100452.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P290341 | BC058990.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P290341 | BC151032.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P290341 | BC151054.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P290341 | AK029927.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P290341 | NM_001100452.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08790807 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127244 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08790069 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03324994 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08790436 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03325364 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08790589 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143497 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08790577 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179181 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08790830 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08790669 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08790762 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109634 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08790757 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03324962 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08790814 | - | - |
| Whole Mouse Genome G4122A | A_52_P122843 | BC058990.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P122843 | AK133487.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P122843 | NM_001100452.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P122843 | AB093219.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P122843 | BC058990.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P122843 | AK133487.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P122843 | NM_001100452.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P122843 | AB093219.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P290341 | BC058990.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P290341 | BC151032.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P290341 | BC151054.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P290341 | AK029927.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P290341 | NM_001100452.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P290341 | BC058990.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P290341 | BC151032.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P290341 | BC151054.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P290341 | AK029927.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P290341 | NM_001100452.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127244 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179181 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109634 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13071191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13185589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13274495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13274497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13516394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13274493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13352962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13129902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13264627 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13121903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13391148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13274491 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13430536 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209153 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10451486 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451491 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451482 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451490 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451488 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451485 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451489 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451484 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451492 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451483 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451487 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440197_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435545_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443567_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443935_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10451481 | - | - |
| Mouse Genome 430 2.0 Array | 1440197_at | - | - |
| Mouse Genome 430 2.0 Array | 1443567_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1443935_at | - | - |
| Mouse Genome 430 2.0 Array | 1443567_at | - | - |
| Mouse Genome 430 2.0 Array | 1435545_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440197_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1443935_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435545_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 210982_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 210982_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700020694.mm.1 | NM_001100452.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020694.mm.1 | NM_001100452.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020694.mm.1 | TC1700002192.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020695.mm.1 | NM_001100452.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020695.mm.1 | NM_001100452.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020695.mm.1 | TC1700002192.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020696.mm.1 | NM_001100452.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020696.mm.1 | NM_001100452.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020696.mm.1 | TC1700002192.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020697.mm.1 | NM_001100452.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020697.mm.1 | NM_001100452.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020697.mm.1 | TC1700002192.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1440197_at | - | - |
| Mouse Expression Set 430 | 1443567_at | - | - |
| Mouse Expression Set 430 | 1435545_at | - | - |
| Mouse Expression Set 430 | 1440197_PM_at | - | - |
| Mouse Expression Set 430 | 1443935_PM_at | - | - |
| Mouse Expression Set 430 | 1443935_at | - | - |
| Mouse Expression Set 430 | 1435545_PM_at | - | - |
| Mouse Expression Set 430 | 1443567_PM_at | - | - |
| Murine Genome U74v2 Set | 167797_r_at | - | - |
| Murine Genome U74v2 Set | 167797_PM_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440197_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443567_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435545_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435545_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443935_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440197_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443567_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443935_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 797059 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2763739 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2763739 | - | - |

Gltscr1l
Genomic Map