Assay Details
| Gene Symbol: Gltscr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100452.1 | NP_001093922.1 | 7 - 8 | 2520 | 65 | |
| GenBank mRNA | AB093219.1 | - | 4 - 5 | 1673 | 65 | |
| AK029927.1 | - | 8 - 9 | 2164 | 65 | ||
| BC032203.1 | - | 4 - 5 | 1201 | 65 | ||
| BC151032.1 | - | 7 - 8 | 2520 | 9087945 | 65 | |
| BC151054.1 | - | 7 - 8 | 2520 | 9087967 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gltscr1l
Genomic Map