Assay Details
| Gene Symbol: Sec24c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168273.1 | NP_001161745.1 | 14 - 15 | 1772 | 63 | |
| NM_172596.2 | NP_766184.2 | 13 - 14 | 2000 | 63 | ||
| GenBank mRNA | AK036519.1 | - | 13 - 14 | 2000 | 63 | |
| AK122199.1 | - | 14 - 15 | 1750 | 63 | ||
| AK132090.1 | - | 8 - 9 | 1027 | 63 | ||
| BC027775.1 | - | 3 - 4 | 375 | 63 | ||
| BC040370.1 | - | 13 - 14 | 1978 | 4239204 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sec24c
Genomic Map