Assay Details
| Gene Symbol: Sec24c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168273.1 | NP_001161745.1 | 21 - 22 | 2739 | 84 | |
| NM_172596.2 | NP_766184.2 | 20 - 21 | 2967 | 84 | ||
| GenBank mRNA | AK036519.1 | - | 20 - 21 | 2970 | 84 | |
| AK122199.1 | - | 21 - 22 | 2717 | 84 | ||
| AK132090.1 | - | 15 - 16 | 1994 | 84 | ||
| BC027157.1 | - | 7 - 8 | 893 | 84 | ||
| BC027775.1 | - | 11 - 12 | 1232 | 84 | ||
| BC040370.1 | - | 20 - 21 | 2945 | 4239204 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sec24c
Genomic Map