| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400001336.mm.1 |
NM_001168273.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001336.mm.1 |
NM_001168273.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001336.mm.1 |
TC1400000170.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400001335.mm.1 |
NM_001168273.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001335.mm.1 |
NM_001168273.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001335.mm.1 |
NM_172596.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001335.mm.1 |
NM_172596.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001335.mm.1 |
TC1400000170.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400001334.mm.1 |
NM_001168273.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001334.mm.1 |
NM_001168273.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001334.mm.1 |
NM_172596.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001334.mm.1 |
NM_172596.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400001334.mm.1 |
TC1400000170.mm.1 |
MTA TC |