Assay Details
| Gene Symbol: Sec24c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168273.1 | NP_001161745.1 | 8 - 9 | 967 | 103 | |
| NM_172596.2 | NP_766184.2 | 7 - 8 | 1195 | 103 | ||
| GenBank mRNA | AK036519.1 | - | 7 - 8 | 1195 | 103 | |
| AK122199.1 | - | 8 - 9 | 945 | 103 | ||
| AK132090.1 | - | 2 - 3 | 222 | 103 | ||
| BC040370.1 | - | 7 - 8 | 1173 | 4239204 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sec24c
Genomic Map