Assay Details
| Gene Symbol: Parva | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020606.5 | NP_065631.3 | 2 - 3 | 485 | 73 | |
| GenBank mRNA | AF237774.1 | - | 2 - 3 | 358 | 73 | |
| AF264766.1 | - | 2 - 3 | 255 | 73 | ||
| AK076473.1 | - | 2 - 3 | 274 | 73 | ||
| AK077368.1 | - | 3 - 4 | 317 | 73 | ||
| AK148878.1 | - | 2 - 3 | 219 | 73 | ||
| AK162796.1 | - | 3 - 4 | 380 | 73 | ||
| BC039750.1 | - | 2 - 3 | 243 | 73 | ||
| BC059236.1 | - | 2 - 3 | 485 | 6400393 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parva
Genomic Map