Assay Details
| Gene Symbol: Parva | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020606.5 | NP_065631.3 | 5 - 6 | 799 | 62 | |
| GenBank mRNA | AB045321.1 | - | 5 - 6 | 672 | 62 | |
| AF237774.1 | - | 5 - 6 | 672 | 62 | ||
| AF264766.1 | - | 5 - 6 | 569 | 62 | ||
| AK076473.1 | - | 5 - 6 | 587 | 62 | ||
| AK077368.1 | - | 6 - 7 | 631 | 62 | ||
| AK148878.1 | - | 5 - 6 | 533 | 62 | ||
| BC039750.1 | - | 5 - 6 | 557 | 62 | ||
| BC059236.1 | - | 5 - 6 | 799 | 6400393 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parva
Genomic Map