Assay Details
| Gene Symbol: Parva | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020606.5 | NP_065631.3 | 9 - 10 | 1061 | 90 | |
| GenBank mRNA | AB045321.1 | - | 9 - 10 | 934 | 90 | |
| AF237774.1 | - | 9 - 10 | 934 | 90 | ||
| AF264766.1 | - | 9 - 10 | 831 | 90 | ||
| AK076473.1 | - | 9 - 10 | 849 | 90 | ||
| AK077368.1 | - | 10 - 11 | 893 | 90 | ||
| AK148878.1 | - | 9 - 10 | 795 | 90 | ||
| BC039750.1 | - | 9 - 10 | 819 | 90 | ||
| BC059236.1 | - | 9 - 10 | 1061 | 6400393 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parva
Genomic Map