Assay Details
| Gene Symbol: Galnt18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173739.3 | NP_776100.2 | 6 - 7 | 1572 | 114 | |
| GenBank mRNA | BC024988.1 | - | 6 - 7 | 1572 | 5044626 | 114 |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt18
Genomic Map