Assay Details
| Gene Symbol: Vcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009502.4 | NP_033528.3 | 7 - 8 | 965 | 85 | |
| GenBank mRNA | AK077850.1 | - | 7 - 8 | 1001 | 85 | |
| AK135530.1 | - | 7 - 8 | 1001 | 85 | ||
| BC008520.1 | - | 7 - 8 | 965 | 3495710 | 85 | |
| L18880.1 | - | 7 - 8 | 894 | 85 | ||
More Information
Set Membership: |
|

Vcl
Genomic Map