Assay Details
| Gene Symbol: Vcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009502.4 | NP_033528.3 | 8 - 9 | 1107 | 81 | |
| GenBank mRNA | AK077850.1 | - | 8 - 9 | 1143 | 81 | |
| BC008520.1 | - | 8 - 9 | 1107 | 3495710 | 81 | |
| L18880.1 | - | 8 - 9 | 1036 | 81 | ||
More Information
Set Membership: |
|

Vcl
Genomic Map