Assay Details
| Gene Symbol: Nfkbie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF030896.1 | - | 1 - 2 | 376 | 94 | |
| AK008843.1 | - | 1 - 2 | 618 | 94 | ||
| AK011965.1 | - | 1 - 2 | 607 | 94 | ||
| AK088585.1 | - | 1 - 2 | 579 | 94 | ||
| AK151450.1 | - | 1 - 2 | 615 | 94 | ||
| AK151692.1 | - | 1 - 2 | 615 | 94 | ||
| AK153248.1 | - | 1 - 2 | 582 | 94 | ||
| BC030923.1 | - | 1 - 2 | 601 | 4909950 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nfkbie
Genomic Map