Assay Details
| Gene Symbol: Nfkbie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF030896.1 | - | 3 - 4 | 701 | 63 | |
| AK008843.1 | - | 3 - 4 | 940 | 63 | ||
| AK011965.1 | - | 3 - 4 | 932 | 63 | ||
| AK088585.1 | - | 2 - 3 | 1514 | 63 | ||
| AK151450.1 | - | 3 - 4 | 940 | 63 | ||
| AK151692.1 | - | 3 - 4 | 940 | 63 | ||
| AK153248.1 | - | 3 - 4 | 907 | 63 | ||
| BC030923.1 | - | 3 - 4 | 926 | 4909950 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nfkbie
Genomic Map