Assay Details
| Gene Symbol: Ttbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162864.1 | NP_001156336.1 | 8 - 9 | 1041 | 65 | |
| GenBank mRNA | AK144536.1 | - | 7 - 8 | 775 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttbk1
Genomic Map