Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081335.2 | NP_001074804.2 | 16 - 17 | 3806 | 89 | |
| GenBank mRNA | AK122351.1 | - | 13 - 14 | 2797 | 89 | |
| AK147675.1 | - | 15 - 16 | 4061 | 89 | ||
| BC026469.1 | - | 1 - 2 | 60 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cul9
Genomic Map