Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081335.2 | NP_001074804.2 | 17 - 18 | 3905 | 98 | |
| GenBank mRNA | AK122351.1 | - | 14 - 15 | 2896 | 98 | |
| AK147675.1 | - | 16 - 17 | 4160 | 98 | ||
| BC026469.1 | - | 2 - 3 | 159 | 98 | ||
More Information
Set Membership: |
|

Cul9
Genomic Map