Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081335.2 | NP_001074804.2 | 19 - 20 | 4187 | 68 | |
| GenBank mRNA | AK039107.1 | - | 1 - 2 | 525 | 68 | |
| AK122351.1 | - | 16 - 17 | 3178 | 68 | ||
| AK147675.1 | - | 18 - 19 | 4442 | 68 | ||
| BC026469.1 | - | 4 - 5 | 441 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cul9
Genomic Map