Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081335.2 | NP_001074804.2 | 20 - 21 | 4314 | 95 | |
| GenBank mRNA | AK122351.1 | - | 17 - 18 | 3305 | 95 | |
| BC026469.1 | - | 5 - 6 | 568 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cul9
Genomic Map