Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081335.2 | NP_001074804.2 | 21 - 22 | 4496 | 67 | |
| GenBank mRNA | AK122351.1 | - | 18 - 19 | 3487 | 67 | |
| AK147675.1 | - | 19 - 20 | 5086 | 67 | ||
| BC026469.1 | - | 6 - 7 | 750 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cul9
Genomic Map