Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081335.2 | NP_001074804.2 | 22 - 23 | 4614 | 55 | |
| GenBank mRNA | AK122351.1 | - | 19 - 20 | 3605 | 55 | |
| AK147675.1 | - | 20 - 21 | 5204 | 55 | ||
| AK162968.1 | - | 1 - 2 | 76 | 55 | ||
| BC026469.1 | - | 7 - 8 | 868 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cul9
Genomic Map