Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081335.2 | NP_001074804.2 | 23 - 24 | 4795 | 63 | |
| GenBank mRNA | AK122351.1 | - | 20 - 21 | 3786 | 63 | |
| AK147675.1 | - | 21 - 22 | 5385 | 63 | ||
| AK162968.1 | - | 2 - 3 | 257 | 63 | ||
| BC026469.1 | - | 8 - 9 | 1049 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cul9
Genomic Map