Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081335.2 | NP_001074804.2 | 26 - 27 | 5376 | 63 | |
| GenBank mRNA | AK051474.1 | - | 3 - 4 | 604 | 63 | |
| AK122351.1 | - | 23 - 24 | 4367 | 63 | ||
| AK147675.1 | - | 24 - 25 | 5966 | 63 | ||
| AK162968.1 | - | 5 - 6 | 836 | 63 | ||
More Information
Set Membership: |
|

Cul9
Genomic Map